Documents
TEM-HC200-AU-100
Sigma-Aldrich
Holey Carbon Film Supported Gold Grids
size 200 mesh, hole size 100 μm, box of 25
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
hole size
100 μm
size
200 mesh
General description
Holey Carbon Film Supported Gold Grids, 200 Mesh, Average hole size is 100 micron, 25/box