Key Documents
TEM-G200CU
Sigma-Aldrich
Glider Grids Standard Copper Square Mesh
size 200 mesh, vial of 100
Synonym(s):
SQUARE 200 MESH COPPER 100/VIAL
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-G200CU is not currently sold in your country. Contact Technical Service
packaging
vial of 100
size
200 mesh
General description
Gilder Grids Standard Square Mesh: Copper Grid, 200 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 125μm, Hole 90μm, Bar 35μm
Choose from one of the most recent versions:
Certificates of Analysis (COA)