Key Documents
TEM-FFGA150-CU-50
Sigma-Aldrich
Formvar Supported Copper Single Hole Grid
hole size 150 μm, box of 50
About This Item
UNSPSC Code:
41111761
packaging
box of 50
hole size
150 μm
General description
A 10 nm thin formvar film supported Single Hole grids. Copper grid, 150 micron hole size. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)