Documents
TEM-CF400AUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Gold Square Mesh
size 400 mesh, box of 50
Synonym(s):
CARBON SQUARE MESH - AU 400 MESH UL 50/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 50
size
400 mesh
General description
A 3-4 nm ultrathin film of pure carbon supported standard squre mesh grids. Gold grid, 400 mesh.