Documents
TEM-CF200F1-AU-TH
Sigma-Aldrich
Carbon Film Supported Gold Gilder Finder F1 Grids
Thick, size 200 mesh, box of 50
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 50
size
200 mesh
General description
A 10 nm thick film of pure carbon supported glider finder (F1) grids. Gold grid, 200 mesh. Gilder finder grids are locator grids with reference/index on the grid: 3.05mm diameter, 0.7 mil (18μm) thickness. F1 type of design has thick bars dividing regions into 6 thin bar areas, which are identified by a numeric system.