Key Documents
TEM-76043-23
Sigma-Aldrich
Cryo-SiN TEM Windows
10 nm Thick, window size 9
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-76043-23 is not currently sold in your country. Contact Technical Service
window size
9
General description
Silicon Nitride 10 nm thick TEM Windows (9 Windows). Ideal substrate for Cryo-Microscopy. Avoid substrate degradation. Plasma cleanable to reduce contaminants.
Choose from one of the most recent versions:
Certificates of Analysis (COA)