Key Documents
TEM-71165-50
Sigma-Aldrich
Cryogenic Grid Box Handling Tool
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-71165-50 is not currently sold in your country. Contact Technical Service
General description
This tool has one end which is threaded and fits into the center hole of the Cryogenic Grid Storage Box (where the screw goes in to secure the lid) for moving the box in and out of the cryogenic chamber.
Choose from one of the most recent versions:
Certificates of Analysis (COA)