Skip to Content
MilliporeSigma
All Photos(1)

Documents

T68802

Sigma-Aldrich

1,2,4-Trimethoxybenzene

97%

Synonym(s):

Hydroxyhydroquinone trimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OCH3)3
CAS Number:
Molecular Weight:
168.19
Beilstein:
2047579
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.533 (lit.)

bp

247 °C (lit.)

density

1.126 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(OC)c(OC)c1

InChI

1S/C9H12O3/c1-10-7-4-5-8(11-2)9(6-7)12-3/h4-6H,1-3H3

InChI key

AGIQIOSHSMJYJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Mihats et al.
International journal of food sciences and nutrition, 68(5), 569-576 (2016-12-06)
Levels of estragole in fennel teas (n = 42) on the Austrian market and the associated dietary exposure were assessed in this study. The estimated daily exposure from consumption of fennel teas ranged from 0.25 to 5.04 μg kg
Elizabeth S Andrews et al.
Journal of chemical ecology, 33(9), 1682-1691 (2007-07-31)
Mutualists and antagonists may place conflicting selection pressures on plant traits. For example, the evolution of floral traits is typically studied in the context of attracting pollinators, but traits may incur fitness costs if they are also attractive to antagonists.
Ting Zhang et al.
Molecules (Basel, Switzerland), 24(24) (2019-12-11)
The unpleasant stale note is a negative factor hindering the consumption of instant ripened Pu-erh tea products. This study focused on investigating volatile chemicals in instant ripened Pu-erh tea that could mask the stale note via sensory evaluation, gas chromatography-mass
Serdar Durdagi et al.
Bioorganic & medicinal chemistry, 19(4), 1381-1389 (2011-02-02)
Carbonic anhydrases (CAs, EC 4.2.1.1) are inhibited by sulfonamides, inorganic anions, phenols, coumarins (acting as prodrugs) and polyamines. A novel class of CA inhibitors (CAIs), interacting with the CA isozymes I, II (cytosolic) and IX, XII (transmembrane, tumor-associated) in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service