Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T64408

Sigma-Aldrich

Gallacetophenone

97%

Synonym(s):

2′,3′,4′-Trihydroxyacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)3C6H2COCH3
CAS Number:
Molecular Weight:
168.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

169-172 °C (lit.)

SMILES string

CC(=O)c1ccc(O)c(O)c1O

InChI

1S/C8H8O4/c1-4(9)5-2-3-6(10)8(12)7(5)11/h2-3,10-12H,1H3

InChI key

XIROXSOOOAZHLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azusa Suzuki et al.
Organic & biomolecular chemistry, 15(37), 7853-7859 (2017-09-08)
A new environmentally responsive fluorescent nucleoside, 3-(naphthalen-1-ylethynyl)-3-deaza-2'-deoxyguanosine (
Sung-Jin Lee et al.
Archives of pharmacal research, 26(10), 832-839 (2003-11-12)
Activated macrophages express inducible isoforms of nitric oxide synthase (iNOS) and cyclooxygenase (COX-2), and produce excessive amounts of nitric oxide (NO) and prostaglandin E2 (PGE2), which play key roles in the processes of inflammation and carcinogenesis. The root of Paeonia
C-Y Oliver Chen et al.
Antioxidants (Basel, Switzerland), 8(4) (2019-04-13)
Almond skins are a waste byproduct of blanched almond production. Polyphenols extracted from almond skins possess antioxidant activities in vitro and in vivo. Thus, we examined the pharmacokinetic profile of almond skin polyphenols (ASP) and their effect on measures of
Ji Young Lee et al.
International journal of molecular sciences, 21(9) (2020-05-06)
Demands for safe depigmentation compounds are constantly increasing in the pharmaceutical and cosmetic industry, since the numerous relevant compounds reported to date have shown undesirable side effects or low anti-melanogenic effects. In this study, we reported three novel inhibitors of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service