Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T35475

Sigma-Aldrich

o-Tolidine

technical grade

Synonym(s):

3,3′-Dimethylbenzidine, 4,4′-Bianisidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-C6H3(CH3)-4-NH2]2
CAS Number:
Molecular Weight:
212.29
Beilstein:
2210640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

mp

128-131 °C (lit.)

SMILES string

Cc1cc(ccc1N)-c2ccc(N)c(C)c2

InChI

1S/C14H16N2/c1-9-7-11(3-5-13(9)15)12-4-6-14(16)10(2)8-12/h3-8H,15-16H2,1-2H3

InChI key

NUIURNJTPRWVAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sensitive colorimetric reagent for gold and for free chlorine in water.

Warning

May be carcinogenic.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service