Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T2032

Sigma-Aldrich

Potassium D-tartrate monobasic

Synonym(s):

D-(−)-Tartaric acid monopotassium salt, D-2,3-Dihydroxybutanedioic acid monopotassium salt, Potassium hydrogen D-tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5KO6
CAS Number:
Molecular Weight:
188.18
Beilstein:
6119984
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

SMILES string

[K+].O[C@@H]([C@H](O)C([O-])=O)C(O)=O

InChI

1S/C4H6O6.K/c5-1(3(7)8)2(6)4(9)10;/h1-2,5-6H,(H,7,8)(H,9,10);/q;+1/p-1/t1-,2-;/m0./s1

InChI key

KYKNRZGSIGMXFH-YGEZSCCGSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Primary standard buffer.

Other Notes

Unnatural isomer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service