Key Documents
SYNV131005
Synthware conical bottom reaction vial with holed compression cap
5 mL, joint: ST/NS 14/10
Synonym(s):
Synthware V131005, reaction tube
Select a Size
1 EA
$25.70
$25.70
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$25.70
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$25.70
Please contact Customer Service for Availability
Joint
joint: ST/NS14/10
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V131005
capacity
5 mL
General description
Supplied with one holed compression cap, one Viton® O-ring and one PTFE -faced silicone rubber septum. For replacement caps, see RC112; For Viton® O-rings, see R014; for septa, see R1420.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Questions
  1. Hello, which magnetic stir bars are compatible with this reaction vial? Thank you
    1 answer
    1. The reaction vial is compatible with a conical stir bar.
      Helpful?