Key Documents
SYNS721240
Synthware high vacuum T-type stopcock with hollow plug
bore size 2 mm, plug size 12.6/40
Synonym(s):
Synthware S721240, T shape stopcock
Select a Size
1 EA
$130.00
$130.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$130.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$130.00
Please contact Customer Service for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S721240
bore size
2 mm
Related Categories
General description
Precision ground with corresponding numbers on the barrel and plug. Lower vacuum chamber is evacuated via a separate 2 mm hole in the plug which is not in the line of flow.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)