Key Documents
SYNM270005
Synthware double manifold with hollow high vacuum stopcocks and SJ 35/20 joint
number of ports, 5, joint: ST/NS 35/20, Connections: front-left-right, rear-right
Synonym(s):
Synthware M270005, vacuum manifold, vacuum unit
Select a Size
1 EA
$959.00
$959.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$959.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
$959.00
Please contact Customer Service for Availability
Joint
joint: ST/NS35/20
feature
number of ports 5
packaging
pkg of 1 ea
manufacturer/tradename
Synthware M270005
General description
With 35/20 ball joints and 4mm double oblique high vacuum hollow glass stopcocks precision ground with corresponding numbers on barrel and plugs. Lower vacuum chambers on stopcock are evacuated with a separate hole in the plug in order to hold the plug in the stopcock barrels.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)