Key Documents
SYNH121410
Synthware Hickman still head with side port, cap, o-ring and PTFE-faced septum
joint: ST/NS 14/10
Synonym(s):
Synthware H121410, Hickman Distillation head
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Joint
joint: ST/NS14/10
manufacturer/tradename
H121410
General description
Synthware Hickman distillation head is designed for microscale fractional distillation. It is supplied with one compression cap, one Viton® O-ring, and one PTFE-faced silicone rubber septum. The side port includes a cap and an 8 mm PTFE-lined septum for syringe injection or sample retrieval.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)