Key Documents
SYNGL32
Synthware Nylon cap with PTFE faced silicone liner
GL 32
Synonym(s):
Synthware GL32, bottle cap
Select a Size
1 EA
$18.90
$18.90
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$18.90
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$18.90
Please contact Customer Service for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware GL32
fitting
GL 32
General description
Replacement caps for use with Solid Phase Reaction Vessels, with PTFE protected Silicone Septum.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)