Documents
SYNF602015F
Synthware Buchner funnel
15 mL, porosity: fine
Synonym(s):
Synthware F602015F, filtering funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F602015F
capacity
15 mL
15 mL
General description
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
Legal Information
Synthware is a trademark of Kemtech America Inc.