Key Documents
SYNF150050
Synthware micro filter flask
capacity 50 mL, stopper size #3
Synonym(s):
Synthware F150050
Select a Size
1 EA
$36.00
$36.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$36.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
$36.00
Please contact Customer Service for Availability
material
flat bottom flask
glass flask
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F150050
capacity
50 mL
General description
Top is tooled for a #2 neoprene filter adapter or #3 rubber stopper.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)