Documents
SYND271420
Synthware distillation distribution adapter
joint: ST/NS 14/20
Synonym(s):
Synthware D271420, short path distillation receiver
About This Item
UNSPSC Code:
41104812
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware D271420
Related Categories
General description
With the outer and inner joint at 135° angle, used with short-path.
Legal Information
Synthware is a trademark of Kemtech America Inc.