Documents
SYNC414250
Synthware Chromatography reservoir with Rodaviss joint
capacity 250 mL, joint: ST/NS 24/40
Synonym(s):
Synthware C414250, glass chromatograpy column
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
colorless
glass
Joint
joint: ST/NS24/40
sterility
non-sterile
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C414250
capacity
250 mL
General description
Reservoir has a top 24/40 or 45/50 Rodaviss outer joint for use with flow controller, and the listed size lower inner joint. Supplied complete with open top cap, Viton® O-ring and loosening ring.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company