Skip to Content
MilliporeSigma
All Photos(1)

Documents

SYNC391740C

Synthware chromatography column with reservoir, coarse fritted disc and PTFE stopcock

250 mL, top joint: ST/NS 24/40, I.D. × L 13.4 mm × 457 mm, coarse fritted disc and PTFE stopcock

Synonym(s):

Synthware C391740C, glass chromatograpy column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NB.51

Joint

joint: ST/NS top24/40

grade

for chromatography

packaging

pkg of 1 ea

manufacturer/tradename

Synthware C391740C

technique(s)

HPLC: suitable

I.D. × L

13.4 mm × 457 mm

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

With a coarse porosity sealed in fritted disc to support column packing. All Synthware columns are heavy wall designed to increase safety.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service