Key Documents
SYNC171200
Synthware water jacketed distillation column with indents to support packing material
joint: ST/NS 14/20, jacket H 200 mm
Synonym(s):
Synthware C171200, jacketed glass distilling column
Select a Size
1 EA
$89.90
$89.90
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$89.90
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
$89.90
Please contact Customer Service for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C171200
jacket H
200 mm
General description
Indentations at the bottom for supporting packing material. Jacket permits the column to be used as a condenser.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)