Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SYNA871420

Synthware 75 degree angled drying tube

joint: ST/NS 14/20

Synonym(s):

Synthware A871420, glass drying tube, Drying tube 95° bent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.

material

colorless tube
glass tube

Joint

joint: ST/NS14/20

packaging

pkg of 1 ea

manufacturer/tradename

Synthware A871420

Looking for similar products? Visit Product Comparison Guide

General description

Angled at 75° to avoid desiccant contact with reactions.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service