Documents
SYNA751420
Synthware 90 degree vacuum adapter with glass stopcock
male joint: ST/NS 14/20
Synonym(s):
Synthware A751420, flow control adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
male joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A751420
Related Categories
General description
Used to regulate the flow of gases or liquids into reaction systems. Stopcock has a bore of 2 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.