Key Documents
SYNA663524
Synthware adapter with top spherical socket joint and lower standard taper inner joint
top joint: ST/NS 35/20, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A663524, adapter with ball socket
Select a Size
1 EA
$52.20
$52.20
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$52.20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$52.20
Please contact Customer Service for Availability
Joint
bottom joint: ST/NS24/40
joint: ST/NS top35/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A663524
General description
Synthware adapter with top spherical socket joint (35/20) and lower standard taper inner joint (24/40) to fit a various of laboratory glassware.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)