Key Documents
SYNA274250
Synthware anti-climb adapter
250 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A274250, rotary evaporator bump trap, bump trap
Select a Size
1 EA
$156.00
$156.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$156.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$156.00
Please contact Customer Service for Availability
Joint
bottom joint: ST/NS24/40
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A274250
capacity
250 mL
Related Categories
General description
Trap provides expanded volume for reactions that have a tendency to foam excessively. The inner vapor tube is sealed to the top of the The inner vapor tube is sealed to the top of thetrap to prevent entry into the condenser.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)