Key Documents
SLWSST108K
PYREX(R) SPARE PTFE KEYS FOR PYREX(R) STOPCOCKS
(straight key bore 8.0 MM)
Synonym(s):
Pyrex, SciLabware SST108K
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Product SLWSST108K is not currently sold in your country. Contact Technical Service
feature
(straight key bore 8.0 MM)
packaging
pkg of 5 ea
manufacturer/tradename
SciLabware SST108K
SciLabware
General description
Stopcocks, plain arms, straight bore, PTFE key
  • Manufactured from borosilicate glass with PTFE keys for highest resistance to chemical attack
  • Barrels are highly polished and the keys are self lubricating, avoiding the need for stopcock grease
  • Keys are fully interchangeable and supplied complete with plastic screw thread retaining device
  • Made to a taper of 1:10 giving a combination of performance and ease of operation
Choose from one of the most recent versions:
Certificates of Analysis (COA)