Documents
SLW2180/02M
Pyrex® Separating funnel, pear-shape, glass key
50 mL, glass stopper, PE stopper, pack of 1 ea
Synonym(s):
Pyrex, SciLabware 2180/02M
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
material
PE stopper
PTFE key
glass stopper
pear-shaped
Joint
joint ST/NS cone19/26
packaging
pack of 1 ea
manufacturer/tradename
SciLabware 2180/02M
funnel diam.
53 mm
stopcock bore
2.0 mm
capacity
50 mL
General description
In liquid-liquid extractions, the two immiscible liquids separate into different layers where the liquid with the higher density collects in the lower part of the vessel and can be drained off via the stopcock.
Features and Benefits
  • Complies with ISO 4800 Type 2
  • Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
  • With interchangeable PTFE stopcock key
  • Supplied with both glass and polyethylene stoppers
  • The 5Litre size is supplementary to the ISO range.
Legal Information
PYREX is a registered trademark of Corning, Inc.