Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SIK7907-10

Sigma-Aldrich

Propylenediphosphonic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H10O6P3
CAS Number:
Molecular Weight:
235.03
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

≥95%

form

solid

shelf life

3 years under the recommended conditions

color

white

InChI

1S/C3H10O6P2/c1-3(11(7,8)9)2-10(4,5)6/h3H,2H2,1H3,(H2,4,5,6)(H2,7,8,9)/p-4/t3-/m0/s1

InChI key

DZQCAOQMXPROIJ-VKHMYHEASA-J

Application

This molecule is used as a crosslinking agent.

Legal Information

Product of SiKEMIA

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service