Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SIK7124-10.4

Sigma-Aldrich

Oleoxein +

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$662.00

$662.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$662.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.23

$662.00


Please contact Customer Service for Availability

Request a Bulk Order

Application

Water and oil repellent treatment for metals, metal oxides and alloys. Solution for nanocoating used to create omniphobic surfaces in electronic devices for passivation.

Legal Information

Product of SiKEMIA

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service