Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S5553

Sigma-Aldrich

Succinimide

98%

Synonym(s):

2,5-Pyrrolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NO2
CAS Number:
Molecular Weight:
99.09
Beilstein:
108440
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

bp

285-290 °C (lit.)

mp

123-125 °C (lit.)

SMILES string

O=C1CCC(=O)N1

InChI

1S/C4H5NO2/c6-3-1-2-4(7)5-3/h1-2H2,(H,5,6,7)

InChI key

KZNICNPSHKQLFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wataru Kamimura et al.
Journal of biomaterials science. Polymer edition, 23(5), 609-628 (2011-02-12)
We have developed a scaffold material consisting of a covalently-bonded structure of alginate and atelocollagen (AtCol). Addition of calcium ions caused the material to form a hydrogel (alginate-modified AtCol gel). The condition of the alginate-modified AtCol gel could be controlled
Mathias Q Müller et al.
Journal of mass spectrometry : JMS, 45(8), 880-891 (2010-07-08)
The fragmentation behavior of a novel thiourea-based cross-linker molecule specifically designed for collision-induced dissociation (CID) MS/MS experiments is described. The development of this cross-linker is part of our ongoing efforts to synthesize novel reagents, which create either characteristic fragment ions
Nada Perisic-Janjic et al.
Journal of pharmaceutical and biomedical analysis, 72, 65-73 (2012-11-14)
Reversed-phase thin-layer chromatographic (RP TLC) retention coefficients for a newly designed series of N-phenyl-3-methyl succinimide derivatives, of a rationally expected anticonvusant activity, were determined as parameters of their lipophilicity. Basic pharmacokinetic descriptors of the agents were calculated in silico with
Zhiwei Lin et al.
Physical chemistry chemical physics : PCCP, 14(30), 10445-10454 (2012-05-05)
Dual-frequency relaxation-assisted two-dimensional infrared (RA 2DIR) spectroscopy was used to investigate energy transport in polyethylene glycol (PEG) oligomers of different length, having 0, 4, 8, and 12 repeating units and end-labeled with azido and succinimide ester moieties (azPEGn). The energy
Ohgi Takahashi et al.
Chemistry & biodiversity, 7(6), 1630-1633 (2010-06-22)
The rapid racemization of aspartic acid (Asp) residues in peptides and proteins is due mainly to the succinimide intermediate. However, there should be another mechanism for Asp racemization without intermediacy of the succinimide. The direct H-atom abstraction from the C(alpha)-atom

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service