Skip to Content
MilliporeSigma
All Photos(1)

Documents

QBD-AQ-11974LF

Sigma-Aldrich

AQuora®550-Maleimide

Synonym(s):

1-(38-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3,31,36-trioxo-7,10,13,16,19,22,25,28-octaoxa-4,32,35-triazaoctatriacontyl)-2-((E)-3-((E)-1-ethyl-3-methyl-5-sulfonato-3-(2,5,8,11-tetraoxatridecan-13-yl)indolin-2-ylidene)prop-1-en-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C75H116N7O28S2
Molecular Weight:
1627.88
UNSPSC Code:
12352111
NACRES:
NA.01

Quality Level

SMILES string

CC(C1=C(N/2CC)C=CC(S(=O)([O-])=O)=C1)(CCOCCOCCOCCOC)C2=C/C=C/C3=[N+](CCC(NCCOCCOCCOCCOCCOCCOCCOCCOCCC(NCCNC(CCN4C(C=CC4=O)=O)=O)=O)=O)C(C=CC(S(=O)([O-])=O)=C5)=C5C3(CCC(NCCOCCOCCOCCOC)=O)C

General description

AQuora® 550-Maleimide is a thiol-reactive fluorophore technology designed to improve solubility during labeling and of the dye labeled conjugate. As a result, the dye labeled conjugates made with AQuora® 550-Maleimide yields enhanced signal and signal-to-noise ratios in fluorescence-based applications including fluorescent western blotting, fluorescence-based microscopy, flow cytometry, and cell-based assays.

This AQuora® 550-Maleimide dye is activated with a maleimide (MAL) group which reacts with a thiol to form a thioether bond.

Legal Information

AQuora is a registered trademark of Quanta BioDesign, Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service