Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD10213

Sigma-Aldrich

Fmoc-N-amido-dPEG®4-acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$202.00
1000 MG
$482.00

$202.00


Please contact Customer Service for Availability


Select a Size

Change View
100 MG
$202.00
1000 MG
$482.00

About This Item

Empirical Formula (Hill Notation):
C26H33NO8
CAS Number:
Molecular Weight:
487.54
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

$202.00


Please contact Customer Service for Availability

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations
reagent type: cross-linking reagent

functional group

Fmoc
amine
carboxylic acid

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

OC(CCOCCOCCOCCOCCNC(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)=O)=O

InChI

1S/C26H33NO8/c28-25(29)9-11-31-13-15-33-17-18-34-16-14-32-12-10-27-26(30)35-19-24-22-7-3-1-5-20(22)21-6-2-4-8-23(21)24/h1-8,24H,9-19H2,(H,27,30)(H,28,29)

InChI key

NUHRPLKTAAVHCZ-UHFFFAOYSA-N

Features and Benefits

Fmoc-N-amido-dPEG®4-acid is a novel Fmoc-protected amino acid containing a single molecular weight PEG (dPEG®) spacer arm. These reagents are used to introduce a hydrophilic, non-immunogenic spacer into a peptide. Three different lengths of dPEG® allow precise control of spacing and hydrophilicity. The Fmoc group can be removed, exposing the terminal amine, using piperidine as with standard peptide synthesis.Fmoc-N-amido-dPEG®n-acid reagents are novel Fmoc-protected amino acids containing a single molecular weight PEG (dPEG®) spacer arm (18.1 Å). This reagents is used to introduce a hydrophilic, non-immunogenic spacer into a peptide. Fmoc group can be removed, exposing the terminal amine, using piperidine as with standard peptide synthesis.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service