Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QBD10170

Sigma-Aldrich

Amino-dPEG®12-OH

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$413.00
1000 MG
$1,350.00

$413.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$413.00
1000 MG
$1,350.00

About This Item

Empirical Formula (Hill Notation):
C24H51NO12
CAS Number:
Molecular Weight:
545.66
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

$413.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCN

InChI

1S/C24H51NO12/c25-1-3-27-5-7-29-9-11-31-13-15-33-17-19-35-21-23-37-24-22-36-20-18-34-16-14-32-12-10-30-8-6-28-4-2-26/h26H,1-25H2

InChI key

JPIQEMLLJLGGFV-UHFFFAOYSA-N

Features and Benefits

Amino-dPEG®12-OH reagent contains a terminal primary amine and a terminal hydroxyl (alcohol) group separated by a single molecular weight dPEG® spacer (42.7 Å). The spacers are highly hydrophilic and non-immunogenic. Because each spacer is a discrete, single molecular weight entity, precise spatial control is possible. These products can be used in a variety of applications, including surface modification where the terminal amine is reacted with an activated carboxylic acid and the terminal hydroxyl provides a non-immunogenic, hydrophilic coating over the targeted surface.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service