Key Documents
PL800GB-7
Plas-Labs butyl glove box gloves
for 1 piece glove set, size 7 (for 8 in. ports)
Select a Size
1 PAIR
$1,170.00
$1,170.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PAIR
$1,170.00
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
$1,170.00
Please contact Customer Service for Availability
material
butyl rubber
size
7 (for 8 in. ports)
feature
reusable
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GB-7
General description
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)