Skip to Content
MilliporeSigma
All Photos(1)

Documents

P71517

Sigma-Aldrich

1-(4-Pyridyl)pyridinium chloride hydrochloride

85%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9ClN2 · HCl
CAS Number:
Molecular Weight:
229.11
Beilstein:
3786515
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

Assay

85%

form

crystals

mp

166-170 °C (dec.) (lit.)

SMILES string

[Cl-].Cl[H].c1cc[n+](cc1)-c2ccncc2

InChI

1S/C10H9N2.2ClH/c1-2-8-12(9-3-1)10-4-6-11-7-5-10;;/h1-9H;2*1H/q+1;;/p-1

InChI key

QZOFFMRDIRXGKJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service