Skip to Content
MilliporeSigma
All Photos(1)

Documents

P26406

Sigma-Aldrich

2-Phenyl-1,3-indandione

97%

Synonym(s):

2-Phenyl-1H-indene-1,3(2H)-dione, Phenindione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O2
CAS Number:
Molecular Weight:
222.24
Beilstein:
1911699
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

crystals

mp

144-148 °C (lit.)

SMILES string

O=C1C(c2ccccc2)C(=O)c3ccccc13

InChI

1S/C15H10O2/c16-14-11-8-4-5-9-12(11)15(17)13(14)10-6-2-1-3-7-10/h1-9,13H

InChI key

NFBAXHOPROOJAW-UHFFFAOYSA-N

Gene Information

human ... VKORC1(79001)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Gelot et al.
Annales de dermatologie et de venereologie, 139(4), 282-286 (2012-04-10)
Antiphospholipid syndrome (APS) is characterised by arterial or venous thrombosis combined with the presence of specific antibodies known as antiphospholipids. It is commonly associated with cutaneous signs. Herein we report a case of atypical cutaneous eruption occurring during the course
Gérard Cam et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 27(4), 1554-1558 (2011-09-21)
Among the vitamin K antagonists (VKA), indanedione-derived VKA is suspected to induce an immunoallergic risk. One indanedione-derived VKA, fluindione, is still being used in France. The aim of this study was to evaluate the contribution of VKA to acute and
Amélie Daveluy et al.
European journal of clinical pharmacology, 68(1), 101-105 (2011-07-28)
Fluindione is an oral vitamin K antagonist (indanedione derivative) exclusively marketed in France and Luxembourg, known to have immuno-allergic adverse effects such as hepatitis, fever or interstitial nephritis. A few cases of drug reaction with eosinophilia and systemic symptoms (DRESS)
C Merklen-Djafri et al.
Annales de dermatologie et de venereologie, 139(3), 199-203 (2012-03-10)
Cutaneous necrosis is a rare complication of vitamin K antagonist therapy. It presents as cutaneous hemorrhagic necrosis and usually occurs at the start of treatment. We describe an atypical case of recurrent skin necrosis after two years of treatment with
"Fulminant" Behçet disease.
Mohamad Zaidan et al.
Presse medicale (Paris, France : 1983), 40(11), 1087-1089 (2011-06-11)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service