Skip to Content
MilliporeSigma
All Photos(2)

Documents

P16656

Sigma-Aldrich

Phenylacetic hydrazide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CONHNH2
CAS Number:
Molecular Weight:
150.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

mp

115-116 °C (lit.)

SMILES string

NNC(=O)Cc1ccccc1

InChI

1S/C8H10N2O/c9-10-8(11)6-7-4-2-1-3-5-7/h1-5H,6,9H2,(H,10,11)

InChI key

FPTCVTJCJMVIDV-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuja Abbas Khan et al.
Pakistan journal of pharmaceutical sciences, 31(4), 1399-1405 (2018-07-24)
The total phenolic content, flavonoid content, in vitro xanthine oxidase (XOD) inhibitory activity and antioxidant activity (AA) of Eucommia ulmoides Oliver leaf extracts were investigated. The AA investigations included 2,2-diphenyl-1-picrylhydrazyl (DPPH) assay, β-carotene/linoleic acid bleaching assay and oxygen radical absorbance
Giuseppe Giannini et al.
Analytical and bioanalytical chemistry, 410(2), 553-564 (2017-11-24)
AvidinOX, the oxidized derivative of Avidin, is a chemically modified glycoprotein, being currently under clinical investigation for targeted delivery of radioactive biotin to inoperable tumors. AvidinOX is produced by 4-hydroxyazobenzene-2-carboxylic acid (HABA)-assisted sodium periodate oxidation of Avidin. The peculiar property
Xiaolai Zhang
Molecules (Basel, Switzerland), 25(2) (2020-01-17)
Benz(o)hydrazide (BH) is the basic aryl hydrazide; aryl hydrazides have been pursued in the course of drug discovery. Oxidations of BH and phenylacetic hydrazide (PAH) by hexachloroiridate(IV) ([IrCl6]2-) were investigated by use of stopped-flow spectral, rapid spectral scan, RP-HPLC and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service