Skip to Content
MilliporeSigma
All Photos(1)

Documents

NCXAUXU100

Sigma-Aldrich

Gold Nanospheres

Ultra Uniform, 100 nm, 0.05 mg/mL (Aqueous 2 mM Sodium Citrate), PEG Carboxyl 0.8 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

nanospheres

concentration

0.05 mg/mL (Aqueous 2 mM Sodium Citrate)

particle size

100 nm

Looking for similar products? Visit Product Comparison Guide

Application

Owing to their plasmonic properties nanoparticles of gold and silver find broad range of applications extending from diagnostics to molecular detection, photothermal and microwave-based therapies, and nanotoxicology. Colloidal silver nanoparticles also find applications in antimicrobial coatings, electronics and photovoltaics, and surface enhanced Raman spectroscopy (SERS) whereas the colloidal gold nanoparticles find applications in lateral flow assays (immunochromatographic assays) and as TEM labels and size standards.

Our ‘Ultra Uniform Gold Nanospheres′ are ideal for applications requiring the highest uniformity in size and shape. These particles have nearly perfect spherical shape, smooth surfaces, and extremely narrow size distributions (CV < 5-6%). These gold nanospheres are available in sizes varying from 10 nm to 100 nm at a concentration of 0.05 mg/mL (gold content) in a 2 mM sodium citrate solution and their surface is stabilized with a carboxyl-terminated ligand.

Due to their high uniformity in size and shape, these Au nanospheres scatter a single color of light under dark field microscope imaging. The high purity of their light scattering signatures makes them perfect scattering labels for imaging and building blocks for plasmonic nanostructures and devices.

Storage and Stability

DO NOT FREEZE. Store at 2-8C when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.

Legal Information

Product of nanoComposix, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service