Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N4207

Sigma-Aldrich

1-Naphthylacetonitrile

97%

Synonym(s):

1-Naphthaleneacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH2CN
CAS Number:
Molecular Weight:
167.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.6192 (lit.)

bp

191-194 °C/18 mmHg (lit.)

mp

33-35 °C (lit.)

SMILES string

N#CCc1cccc2ccccc12

InChI

1S/C12H9N/c13-9-8-11-6-3-5-10-4-1-2-7-12(10)11/h1-7H,8H2

InChI key

OQRMWUNUKVUHQO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alina Plenis et al.
Chromatographia, 76(5-6), 255-265 (2013-03-14)
A new liquid chromatographic (LC) method for simultaneous determination of lidocaine hydrochloride (LH) and tribenoside (TR) along with their related compounds in pharmaceutical preparations is described. Satisfactory LC separation of all analytes after the liquid-liquid extraction (LLE) procedure with ethanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service