Skip to Content
MilliporeSigma
All Photos(1)

Documents

N25001

Sigma-Aldrich

N-Nitrosodimethylamine

reagent grade

Synonym(s):

Dimethylnitrosamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NNO
CAS Number:
Molecular Weight:
74.08
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

reagent grade

vapor pressure

5 mmHg ( 20 °C)

refractive index

n20/D 1.437 (lit.)

bp

153 °C/774 mmHg (lit.)

density

1.01 g/mL (lit.)

SMILES string

CN(C)N=O

InChI

1S/C2H6N2O/c1-4(2)3-5/h1-2H3

InChI key

UMFJAHHVKNCGLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Induces gastric, liver, kidney and lung cancer in mice and rats.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Carc. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Iwaniuk et al.
Human & experimental toxicology, 34(6), 591-600 (2014-10-12)
N-nitrosodimethylamine (NDMA) is a xenobiotic widespread in human environment capable of regulating the lifespan of immune cells. In this study, we examined the roles of the tumor necrosis factor-related apoptosis-inducing ligand (TRAIL)/death receptor 5 (DR5) complex and the Fas molecule

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service