Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N22657

Sigma-Aldrich

4-Nitrophenyl trifluoroacetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CO2C6H4NO2
CAS Number:
Molecular Weight:
235.12
Beilstein:
658785
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

bp

120 °C/12 mmHg (lit.)

mp

35-39 °C (lit.)

application(s)

peptide synthesis

SMILES string

[O-][N+](=O)c1ccc(OC(=O)C(F)(F)F)cc1

InChI

1S/C8H4F3NO4/c9-8(10,11)7(13)16-6-3-1-5(2-4-6)12(14)15/h1-4H

InChI key

JFOIBTLTZWOAIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service