Skip to Content
MilliporeSigma
All Photos(1)

Documents

N21901

Sigma-Aldrich

4-Nitrophenyl phenyl ether

98%

Synonym(s):

1-Nitro-4-phenoxybenzene, 4-Nitrodiphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OC6H5
CAS Number:
Molecular Weight:
215.20
Beilstein:
2051911
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

bp

320 °C (lit.)

mp

53-56 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Oc2ccccc2)cc1

InChI

1S/C12H9NO3/c14-13(15)10-6-8-12(9-7-10)16-11-4-2-1-3-5-11/h1-9H

InChI key

JDTMUJBWSGNMGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Miyauchi et al.
Chemico-biological interactions, 51(1), 49-62 (1984-09-01)
A series of possible metabolites--4-nitrosobiphenyl ether (4-NO), 4-hydroxylaminobiphenyl ether (4-NHOH), 4-aminobiphenyl ether (4-NH2), 4-hydroxyacetylaminobiphenyl ether (4-N(OH)Ac), 4-acetoxyacetylaminobiphenyl ether (4-N(OAc)Ac)involved in the toxic effects of 4-nitrobiphenyl ether (4-NO2) was synthesized and tested for mutagenic activity toward Salmonella typhimurium TA100 strain in
R L Gupta et al.
Toxicology letters, 34(1), 13-21 (1986-11-01)
The mutagenic potential of 4-nitrodiphenyl ether (NDE) an intermediate of herbicides and an anthelmintic drug, and its reduced products (-NO, -NHOH) were investigated in the Ames test. All the three compounds were potent direct-acting base-pair and frameshift mutagens with activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service