Key Documents
MT59904377
Mettler Toledo RedCap ORP probe, Redox indicating electrode
glass shaft, platinum ring, L × W 120 mm × 12 mm
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
material
glass shaft
description
S7 connection
Without cable
manufacturer/tradename
Mettler-Toledo 59904377
L × W
120 mm × 12 mm
General description
PT805-S7/120