Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MT51350108

Mettler-Toledo ISA solution

0.9 mol/L Aluminum Sulfate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51191600
NACRES:
NB.79

form

liquid

manufacturer/tradename

Mettler-Toledo 51350108

volume

250 mL

Looking for similar products? Visit Product Comparison Guide

General description

An ISA solution is used to level the ion activity for all sample solutions to enable ISE measurements in sample solutions. If the sample solution needs additional removal of interfering ions and/or pH buffering it is not called ISA solution, but TISAB solution.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service