Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MT51344750

Mettler Toledo perfectION reference filling solution A

volume 5 x 60 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51191600
NACRES:
NB.79
Pricing and availability is not currently available.

form

liquid

manufacturer/tradename

Mettler-Toledo 51344750

volume

5 x 60 mL

Legal Information

perfectION is a trademark of Mettler-Toledo, Inc.

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Ox. Liq. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service