Skip to Content
MilliporeSigma
All Photos(1)

Documents

MT51343191

Mettler-Toledo InLab® Reference Pro electrode

Reference electrode with gel electrolyte and movable PTFE-sleeve

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
39121436
NACRES:
NB.79

material

glass shaft

description

Reference electrolyte Gel
Reference system Ag/AgCl
S7 connector
Without cable

manufacturer/tradename

Mettler-Toledo 51343191

temperature measuring range

0-60 °C

L × diam.

120 mm × 12 mm

General description

Mettler Toledo InLab® Reference Pro is a special reference electrode with glass body and S7 screw head.
The InLab® Reference Pro is the perfect reference electrode for measurements in ion-selective half-cells. For metal or ISE half-cells
  • Movable PTFE-sleeve
  • Bridge electrolyte 3 mol/L KCl

Legal Information

InLab is a registered trademark of Mettler-Toledo, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service