Skip to Content
MilliporeSigma
All Photos(1)

Documents

MT51343157

Mettler-Toledo InLab® Surface pH electrode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115604
NACRES:
NB.61

material

polysulfone shaft

description

Reference electrolyte 3 mol/L KCl satur. with AgCl
Reference system Ag/AgCl
S7 connector
without cable

manufacturer/tradename

Mettler-Toledo 51343157

temperature measuring range

0-50 °C

L × diam.

110 mm × 12 mm

pH range

1-11

General description

InLab® Surface is a combination pH electrode with flat membrane for measuring samples such as skin, leather, paper and agar.
Polysulphone body and S7 screw head.

Designed for measuring samples with a flat surface, the InLab® Surface has a rugged polysulphone shaft for added protection in hostile environments.

Combination design avoids use of separate reference and indicating electrodes. The innovative Safelock closing ring gives easy access for re-filling reference electrolyte and complete sealing when closed. Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
  • Flat membrane
  • Combination design
  • Safelock closing ring
  • Detachable cable
  • Rugged polysulphone shaft

Legal Information

InLab is a registered trademark of Mettler-Toledo, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service