Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MT51340345

Mettler-Toledo Economic & Easy pH electrode

DIN connection, glass, HA membrane, glass shaft, Suitable for TRIS buffers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115604
NACRES:
NB.79

material

glass shaft
glass, HA membrane

description

Fill solution: 3M KCl sat. AgCl
Reference system: Ag/AgCl

feature

Suitable for TRIS buffers

manufacturer/tradename

Mettler-Toledo 51340346

temperature measuring range

0-80 °C

L × diam.

120 mm × 12 mm

pH range

0-14

Looking for similar products? Visit Product Comparison Guide

General description

The LightEdition (LE) sensors are tailor-made for high price/performance ratio. The offering includes sensors for all common applications and focuses on low maintenance and easy handling.

It is the perfect match with the Five and Education Line meters.
Supplied with 1.0 m cable.
pH electrode LE420-DIN
The movable PTFE sleeve junction makes the LE420 the right choice for emulsions, TRIS buffers and any other challenging application.
Movable PTFE reference junction
Suitable for low hydrogen concentration

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service