Skip to Content
MilliporeSigma
All Photos(1)

Documents

MT51325005

Mettler-Toledo density standards

set of 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115312
NACRES:
NB.61

packaging

set of 10

manufacturer/tradename

Mettler-Toledo 51325005

General description

For use with Mettler Toledo Density meter (MT51324450)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service